SHOWA™ Natural Rubber Latex HD™ Gloves
Two-color wear indicator
Manufacturer: SHOWA™ 55811
 View more versions of this product
Catalog No. 19-042-656
  Check Availability
Add to Cart
    Specifications
    Color Black, red
    Exterior Finish Bisque
    Glove Cuff Style Rolled
    For Use With (Application) manufacturing, aircraft assembly, janitorial, plant maintenance, agriculture, heavy duty plant maintenance, chemical processing, sanitization clean up, sandblasting/ electroplating
    Length (English) 18 in.
    Length (Metric) 45.7cm
    Material Natural Rubber
    Powdered Yes
    Size 2X-Large
    Sterile No
    Thickness 40mil
    Certifications 21 CFR 170-199, EN 388 1120, CE
    Heavy-duty gloves are ideal for dip tank, sandblasting, and electroplating applications.
    • Acid- and abrasion-resistant exterior features
    • Bisque finish for superior gripping
    • Inside of gloves is smooth and unflocked
    • Powdered interior and rolled-edge cuffs expedite donning and doffing
    • Two-color composition (black exterior/red interior) serves as a visual wear indicator
    • Hand specific, nonsterile, and liquidproof
    Gloves are not intended for electrical use.
  • Description & Specifications
Related Products
Product Content Feedback