Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 6797 Chemical Resistant Neoprene Gloves, Extended Sleeve
Click to view available options
16 in. (41 cm) Long; Smooth
Description
- Extended-arm protection
- Protection against acids, caustics, oils, greases and many solvents
- Protection against physical hazards such as abrasion and cut
- Cotton liner is cool and comfortable in hot conditions and keeps wearer warm in cold conditions
- Ideal applications include:
- Chemical spray and treatment
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
Specifications
Certifications/Compliance | 21 CFR 170-199 |
Certifications/Compliance (Europe) | Cat III CE 0321, EN 407:3242XX |
Color | Black |
Latex Free | Yes |
Material | Neoprene |
Size | 10 |
EN388 Standard | EN 388:3111X |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | AJKLMT |
EN ISO 374 - 5 | Bacteria, Fungi only |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction