Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA Neoprene Gloves
Wet-grip neoprene glove
$87.00 - $138.00
Specifications
Certifications/Compliance | 21 CFR 170-199 |
---|---|
For Use With (Application) | manufacturing, refining, Petrochemicals, degreasing, plating, handling Oils, Acids, Caustics, Alcohols and Solvents |
Glove Cuff Style | Gauntlet |
Lining | Five-piece Cotton |
Color | Black |
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
19-017980
|
SHOWA
678010 |
12 in. |
Case of 72 for $87.00
|
|
19-018116
|
SHOWA
678410 |
14 in. |
Dozen of 12 for $112.00
Case of 3 DZ for $336.00 |
|
19-018119
|
SHOWA
679710 |
18 in. |
Case of 36 for $138.00
|
|
Description
Provide protection against acids, caustics, oils, greases and many solvents.
- Multi-dipped in premium-grade neoprene for superior quality and durability
- Protect against physical hazards such as abrasion and cut
- Cotton liner
- Sanitized
- Color: Black
Specifications
21 CFR 170-199 | |
Gauntlet | |
Black | |
10 |
manufacturing, refining, Petrochemicals, degreasing, plating, handling Oils, Acids, Caustics, Alcohols and Solvents | |
Five-piece Cotton | |
Neoprene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title