SHOWA™ Nitri-Flex™ Kevlar™ Gloves

Flexible nitrile-dipped gloves with Kevlar™ liner provide comfort with durability

Manufacturer: SHOWA™ 490010

 View more versions of this product

Catalog No. 19-050-303D

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Weight Medium weight
    Glove Cuff Style Knit
    Hand Hand-specific
    For Use With (Application) Shearing, Metal fabrication and assembly, general construction, facilities maintenance, warehouse and shipping jobs
    Length (English) 10 in.
    Length (Metric) 25.5cm
    Material Kevlar™
    Size X-Large
    Coating Material Textured
    Color Yellow, green
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit