Learn More
SHOWA™ NRK™ Cut-Resistant Gloves Available on GSA/VA Contract for Federal Government customers only.
Provide excellent cut and slip resistance and dexterity
Manufacturer:  SHOWA™ NRK07
 View more versions of this product
Catalog No. 19-050013A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 100% Kevlar™ with a screen-printed coating
  • Thickness: 10-gauge,
  • Seamless design
Not designed for use with moving or serrated blades.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Safety
Level 2
Dupont™ Kevlar™ Aramid Fiber
Yellow
PVC
Material handling, mechanical and automotive, machine or compartment assembly, parts assembly, Glass handling
Small
Yellow