Learn More
SHOWA™ NRK™ Cut-Resistant Gloves
Provide excellent cut and slip resistance and dexterity
Manufacturer:  SHOWA™ NRK08
 View more versions of this product
Catalog No. 19-050013B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 100% Kevlar™ with a screen-printed coating
  • Thickness: 10-gauge,
  • Seamless design
Not designed for use with moving or serrated blades.
Specifications
Specifications
Safety
Level 2
Dupont™ Kevlar™ Aramid Fiber
Yellow
PVC
Material handling, mechanical and automotive, machine or compartment assembly, parts assembly, Glass handling
Medium
Yellow