Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ NRK™ Cut-Resistant Gloves
Click to view available options
Size:
Small
Medium
X-Large
Description
- Aramid fiber with a screen-printed coating
- Thickness: 10-gauge,
- Seamless design
Not designed for use with moving or serrated blades.
Specifications
Coating Material | PVC |
Color | Yellow |
Cut Resistance Rating | ANSI A2 |
Material | Aramid Fiber |
Product Type | Cut-resistant Gloves |
Size | Small |
For Use With (Application) | Material handling, mechanical and automotive, machine or compartment assembly, parts assembly, Glass handling |
Glove Cuff Color | Yellow |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction