SHOWA™ Strapper™ Gloves

Excellent replacement for leather driver's style glove

Manufacturer: SHOWA™ 279010

 View more versions of this product

Catalog No. 19-014540

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Textured
    Glove Cuff Style Slip-on
    For Use With (Application) assembly, inspection, light-to-medium fabrication and material handling
    Material Nitrile
    Liner Material Cotton
    Size 10
    Sterile Yes
    Coated Area Palm
    Color Navy
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit