Learn More
SHOWA™ Super Flex™ Gloves
Low-temperature-resistant PVC formulation soft and supple even in cold conditions
Manufacturer:  SHOWA™ 75610
 View more versions of this product
Catalog No. 19-056747
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Provide excellent abrasion resistance and protection from a broad range of chemicals.
  • Cotton jersey liner enhances worker comfort and glove flexibility
  • Yellow color provides a high-contrast, visual warning of possible contamination
  • Fully coated
  • Smooth finish
  • Sanitized™
Specifications
Specifications
Safety
Smooth
manufacturing, light duty maintenance, refining, Petrochemicals, mining, construction, utilities
Cotton Jersey
Yellow
18 in.
Full
Gauntlet
PVC
10
Sterile
45.7 cm