SHOWA™ Value Master™ Natural Rubber Gloves
Resist animal fats, caustics, acids, oils, salts, alcohols, and detergents
Manufacturer: SHOWA™ VMUM08
 View more versions of this product
Catalog No. 19-019707
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 170-199, CE
    Exterior Finish Tractor tread
    Glove Cuff Style Rolled
    For Use With (Application) manufacturing, aircraft assembly, janitorial, plant maintenance, food processing, poultry processing, sanitization clean up, household cleaning
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Natural Rubber
    Size Medium
    Thickness 18mil
    Color Amber
  • Description & Specifications
Related Products
Product Content Feedback