Learn More
SHOWA™ Value Master™ Natural Rubber Gloves
Resist animal fats, caustics, acids, oils, salts, alcohols, and detergents
Manufacturer:  SHOWA™ VMUL09
 View more versions of this product
Catalog No. 19-019720
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tractor tread
manufacturing, aircraft assembly, janitorial, plant maintenance, food processing, poultry processing, sanitization clean up, household cleaning
Large
21 CFR 170-199, CE
12 in.
Rolled
Rubber, natural
18mil
Amber
30.5cm