Learn More
SHOWA™ Value Master™ Natural Rubber Gloves
Resist animal fats, caustics, acids, oils, salts, alcohols, and detergents
Manufacturer:  SHOWA™ VM07
 View more versions of this product
Catalog No. 11-395-23A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tractor tread
manufacturing, aircraft assembly, janitorial, plant maintenance, food processing, poultry processing, sanitization clean up, household cleaning
Rubber, natural
18mil
Yellow
30.5cm
Rolled
Flock
Small
21 CFR 170-199, CE
12 in.