Learn More
beta-2 Adrenergic R/ADRB2 Mouse anti-Human, Alexa Fluor™ 350, Clone: 586107, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB100401U100UG
Catalog No. FB10401U100
Add to cart
This item is not returnable. View return policy
Description
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
beta-2 Adrenergic R/ADRB2
586107
Flow Cytometry 0.25-1 μg/106 cells
P07550
ADRB2, ADRB2R, ADRBR, adrenergic, beta-2-, receptor, surface, B2AR, B2ARADRB2R, BAR, beta-2 adrenergic receptor, beta2 AdrenergicR, beta-2 AdrenergicR, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
NS0 mouse myeloma cell line transfected with human beta-2 Adrenergic R/ADRB2, Accession # P07550
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human beta-2 Adrenergic R/ADRB2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
154
Human