Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ beta 2-Microglobulin Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796575PLATES

 View more versions of this product

Catalog No. NB010354


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat beta 2-Microglobulin.
Specifications

Specifications

HRP
Antibody Pairs
2.66 pg/mL
567
RUO
beta 2-Microglobulin
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Rat
B2M
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 170 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat beta 2-Microglobulin.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-beta 2-Microglobulin monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Rat beta 2-Microglobulin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 35 ng of recombinant Rat beta 2-Microglobulin.
  • Storage is content dependent.
    Rabbit
    5 Plates
    2.66-170 pg/mL
    beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit