Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ beta 2-Microglobulin Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$832.00
Specifications
Assay Range | 3.13 pg/mL |
---|---|
Assay Sensitivity | 3.13-200 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009797
|
Novus Biologicals™
NBP27937915PLATES |
15 Plates |
Each of 1 for $832.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human beta 2-Microglobulin.Specifications
3.13 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
B2M | |
RUO | |
Storage is content dependent. |
3.13-200 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human beta 2-Microglobulin. | |
Sandwich ELISA | |
567 | |
Mouse | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
beta 2-Microglobulin |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title