Learn More
beta 2-Microglobulin Mouse anti-Human, DyLight 680, Clone: 3G5H8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237287FR
Catalog No. NBP237287FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
beta 2-Microglobulin
3G5H8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
B2M
Purified recombinant fragment of human B2M (AA: 21-100) expressed in E. Coli.
Protein A or G purified
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary
567
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human