Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Mouse anti-Human, PerCP, Clone: 3G5H8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237287PCP

Catalog No. NBP237287W

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

beta 2-Microglobulin
3G5H8
Flow Cytometry
Purified
B2M
Purified recombinant fragment of human B2M (AA: 21-100) expressed in E. Coli.
Protein A or G purified
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary
567
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit