Learn More
beta 2-Microglobulin Mouse anti-Rat, Alexa Fluor 405, Clone: 747502, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3864V100UG
Catalog No. FAB3864V100
Add to cart
This item is not returnable. View return policy
Description
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
beta 2-Microglobulin
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P07151
B2M
Mouse myeloma cell line NS0-derived recombinant rat beta 2-Microglobulin Ile21-Met119 Accession # P07151
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects rat beta 2-Microglobulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat alpha 2-Macroglobulin is observed.
Flow Cytometry
747502
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
B2M, beta 2Microglobulin, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Mouse
IgM
RUO
Primary
567
Rat
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)