Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Rabbit anti-Human, Clone: 201, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | beta 2-Microglobulin |
---|---|
Clone | 201 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
beta 2-Microglobulin | |
Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Recombinant Human beta 2-Microglobulin protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
201 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
0.2 um filtered solution in PBS | |
567 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title