Learn More
beta-3 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P2411MG

Description
AA Sequence of recombinant protein: GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-3. Anti-Human BD-3-specific antibody was purified by affinity chromatography employing an immobilized Human BD-3 matrix. Sandwich ELISA: To detect Human BD-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-3 (500-P241Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-3. Western Blot: To detect Human BD-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
beta-3 Defensin Polyclonal antibody specifically detects beta-3 Defensin in Human samples. It is validated for ELISA, Western BlotSpecifications
beta-3 Defensin | |
Polyclonal | |
PBS with no preservative | |
P81534 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
414325 | |
-20°C |
ELISA, Western Blot | |
Unconjugated | |
DEFB103A | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
E.coli-derived Recombinant Human BD-3 | |
1 mg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.