Learn More
beta-3 Tubulin Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ARC0456, Invitrogen™
Rabbit Monoclonal Antibody
Manufacturer:  Invitrogen MA535189
Catalog No. PIMA535189
Add to cart
Description
Description
The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.
Specifications
Specifications
beta-3 Tubulin
ARC0456
PBS with 50% glycerol and 0.2% sodium azide; pH 7.3
Q13509, Q4QRB4, Q9ERD7
3200002H15Rik; beta-4; Beta-tubulin class-III; BOS_17166; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; I79_000194; M(beta)3; M(beta)6; MC1R; Neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
10381, 22152, 246118
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
TUBB3
Liquid
TUBB3
A synthesized peptide derived from human beta Tubulin III
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat