Learn More
Human beta-3 Adrenergic R/ADRB3 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB9419U100UG
Catalog No. FAB9419U100
Add to Cart
This item is not returnable. View return policy
Description
Description
beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
beta-3AdrenergicR/ADRB3
Monoclonal
Alexa Fluor 350
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945
100 μg
155
Human
Supernatant
Flow Cytometry
493324
Flow Cytometry 0.25-1μg/106 cells
P13945
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Product Suggestions
Product Suggestions