Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-3 Adrenergic R/ADRB3 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9419S100UG
This item is not returnable.
View return policy
Description
beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry.Specifications
beta-3AdrenergicR/ADRB3 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945 | |
100 μg | |
155 | |
Human | |
Supernatant |
Flow Cytometry | |
493324 | |
Flow Cytometry 0.25-1μg/106 cells | |
P13945 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction