Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR5 Antibody (85B152.5) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 26 publications

$181.90 - $487.50

Specifications

Antigen TLR5
Clone 85B152.5
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells in 100 ul, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 17570691), Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 24779433), Flow (Intracellular) reported in scientific literature (PMID 24779433)
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP197728SS
SDP
View Documents
Novus Biologicals
NBP197728SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP197728
SDP
View Documents
Novus Biologicals
NBP197728
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).
Specifications

Specifications

TLR5
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
O60602
7100
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
85B152.5
Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells in 100 ul, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 17570691), Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 24779433), Flow (Intracellular) reported in scientific literature (PMID 24779433)
Monoclonal
Purified
RUO
Human, Mouse, Canine
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
TLR5
IgG2a
Protein G purified
100 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.