Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ beta-Actin Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

$230.50

Specifications

Product Type Recombinant Protein Antigen
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
Gene Symbol ACTB
View More Specs

Products 2
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
NBP254690X View Documents Novus Biologicals™
NBP254690PEP
PKANREKMTQIMFETFNTPAMYVAIQ Each for $230.50
Add to cart
 
NBP254691X View Documents Novus Biologicals™
NBP254691PEP
MVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIVTNWD Each for $230.50
Add to cart
 
Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human beta-Actin The beta-Actin Recombinant Protein Antigen is derived from E. coli. The beta-Actin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

Recombinant Protein Antigen
RUO
ACTB
PBS and 1M Urea, pH 7.4
E.coli
Unlabeled
Yes
Store at −20°C. Avoid freeze-thaw cycles.
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Human
AC
22 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit