Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ beta-Alaninamide hydrochloride, 96%

Click to view available options
Quantity:
5g
25g
100g
64017-81-8
C3H9ClN2O
124.568
MFCD00060747
GAGJMOQGABUOBK-UHFFFAOYSA-N
3-aminopropanamide hydrochloride,3-aminopropionamide hydrochloride,3-aminopropanamide hcl,beta-alaninamide hydrochloride,h-beta-ala-nh2.hcl,h-beta-ala-nh2 hcl,3-amino-propionamide hydrochloride,h-ala-nh2hcl,propanamide, 3-amino-, monohydrochloride,h-beta-ala-nh2hcl
22222288
3-aminopropanamide;hydrochloride
C(CN)C(=O)N.Cl
64017-81-8
C3H9ClN2O
124.568
MFCD00060747
GAGJMOQGABUOBK-UHFFFAOYSA-N
3-aminopropanamide hydrochloride,3-aminopropionamide hydrochloride,3-aminopropanamide hcl,beta-alaninamide hydrochloride,h-beta-ala-nh2.hcl,h-beta-ala-nh2 hcl,3-amino-propionamide hydrochloride,h-ala-nh2hcl,propanamide, 3-amino-, monohydrochloride,h-beta-ala-nh2hcl
22222288
3-aminopropanamide;hydrochloride
C(CN)C(=O)N.Cl
Applications
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
Beilstein 3909917
Formula Weight 124.57
Physical Form Solid
Percent Purity 96%
Melting Point 149°C to 154°C
Quantity 25g
Chemical Name or Material beta-Alaninamide hydrochloride

storageNote1 : 2° to 8°C