Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ beta-Alanine tert-butyl ester hydrochloride, 95%

Click to view available options
Quantity:
5g
25g
100g
58620-93-2
C7H16ClNO2
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride,h-beta-ala-otbu.hcl,beta-alanine tert-butyl ester hydrochloride,h-beta-ala-otbu hcl,beta-alanine t-butyl ester hydrochloride,3-amino-propionic acid tert-butyl ester hydrochloride,h-?-ala-otbu.hcl,h-,a-ala-otbu.hcl,h-beta-ala-otbu hydrochloride,tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
CC(C)(C)OC(=O)CCN.Cl
58620-93-2
C7H16ClNO2
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride,h-beta-ala-otbu.hcl,beta-alanine tert-butyl ester hydrochloride,h-beta-ala-otbu hcl,beta-alanine t-butyl ester hydrochloride,3-amino-propionic acid tert-butyl ester hydrochloride,h-?-ala-otbu.hcl,h-,a-ala-otbu.hcl,h-beta-ala-otbu hydrochloride,tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
CC(C)(C)OC(=O)CCN.Cl
Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Linear Formula H2NCH2CH2COOC(CH3)3·HCl
Beilstein 6660122
Formula Weight 181.66
Physical Form Solid
Percent Purity 95%
Melting Point 167°C to 169°C
Quantity 25g
Chemical Name or Material beta-Alanine tert-butyl ester hydrochloride

storageNote1 : -30° to -10°C