Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ beta-Alanyl-L-histidine, 95%

Click to view available options
Quantity:
5g
25g
305-84-0
C9H14N4O3
226.236
MFCD00005207
CQOVPNPJLQNMDC-ZETCQYMHSA-N
l-carnosine,carnosine,beta-alanyl-l-histidine,karnozin,karnozzn,ignotine,l-ignotine,n-2-m,unii-8ho6pvn24w,nalpha-beta-alanyl-l-histidine
439224
CHEBI:15727
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
305-84-0
C9H14N4O3
226.236
MFCD00005207
CQOVPNPJLQNMDC-ZETCQYMHSA-N
l-carnosine,carnosine,beta-alanyl-l-histidine,karnozin,karnozzn,ignotine,l-ignotine,n-2-m,unii-8ho6pvn24w,nalpha-beta-alanyl-l-histidine
439224
CHEBI:15727
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
Beilstein 87671
Formula Weight 226.24
Physical Form Crystalline
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material beta-Alanyl-L-histidine

RTECSNumber : MS3080000

TSCA : TSCA

storageNote1 : -30° to -10°C