Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid Monoclonal Antibody (DE2B4), HRP, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA546599

Encompass

Catalog No. PIMA546599


Add to Cart
This item is not returnable. View return policy

Description

Description

beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Bovine, Human, Porcine samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta Amyloid
Monoclonal
1 mg/mL
PBS with 0.01% thimerosal
P05067, P79307, Q28053
APP
Synthetic peptide corresponding to residues 1 - 16 of the human amyloid beta protein
50 μg
Primary
Bovine, Human, Porcine
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation
DE2B4
HRP
APP
A4; AAA; ABETA; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; amyloid beta (A4) precursor protein (peptidase nexin-II, Alzheimer disease); amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; amyloid precursor protein variant 1; amyloid precursor protein variant 2; Amyloid β; amyloid-beta A4 protein; amyloid-beta A4 protein; amyloid beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; beta amyloid protein; beta amyloid protein precursor; beta-amyloid peptide; be
Mouse
Affinity chromatography
RUO
280722, 351, 397663
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit