Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid Mouse anti-Human, Alexa Fluor™ 594, Clone: 16000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223544AF594

Catalog No. NP22344A94


Add to cart

Description

Description

beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for ELISA.
Specifications

Specifications

beta Amyloid
16000000000
ELISA
Conjugated, Purified
APP
Amyloid Beta-Peptide (5-16) adsorbed onto aluminum hydroxide gel
Protein A or G purified
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary
351.0
Human
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Abeta, Amyloid beta, beta-amyloid peptide
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
The binding of Clone 16E9 to amyloid Beta-peptide depends on the presence of at least amino acid residues 10-16.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit