Learn More
beta Amyloid Mouse anti-Human, Alexa Fluor™ 750, Clone: 16000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223544AF750
Catalog No. NP22344A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for ELISA.
Specifications
Specifications
beta Amyloid
16000000000
ELISA
Abeta, Amyloid beta, beta-amyloid peptide
Mouse
IgG2a Kappa
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary
351
Human
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
APP
Amyloid Beta-Peptide (5-16) adsorbed onto aluminum hydroxide gel
0.1 mL
Store at 4°C in the dark.
Monoclonal
The binding of beta Amyloid antibody (16E9) to amyloid Beta-peptide depends on the presence of at least amino acid residues 10-16.