Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Arrestin 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP258092

Catalog No. NBP258092


Add to cart

Description

Description

beta-Arrestin 1 Polyclonal antibody specifically detects beta-Arrestin 1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

beta-Arrestin 1
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
ARB1, ARR1, arrestin 2, Arrestin beta-1, arrestin, beta 1, beta-arrestin-1
Rabbit
IgG
100 ul
Signal Transduction, Virology Bacteria and Parasites
Primary
408
Human
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
ARRB1
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PKPKEEPPHREVPENETPVDTNL
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only