Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Arrestin 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | beta-Arrestin 2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.25 - 0.5 ug/mL, Simple Western 1:100, Immunohistochemistry-Paraffin 5 ug/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224569SS
![]() |
Novus Biologicals
NBP224569SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224569
![]() |
Novus Biologicals
NBP224569 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
beta-Arrestin 2 Polyclonal specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
beta-Arrestin 2 | |
Western Blot 0.25 - 0.5 ug/mL, Simple Western 1:100, Immunohistochemistry-Paraffin 5 ug/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Xenopus species, Zebrafish | |
ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
ARRB2 | |
IgG | |
Affinity Purified | |
The eIF3A amino acid sequence used as immunogen for the antibody is 100% homologous in human, rat, mouse, Xenopus, chicken, chimpanzee, zebrafish, cow, platypus, horse, opossum and boar, and 94% homologous in Tetradon. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
409 | |
A peptide sequence corresponding to an area between amino acids 1 and 50 of human ARRB2 was used as the immunogen for the antibody, Gen Bank no. gb:ABG47460.1. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title