Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ beta-Bag Cell Peptide
For cellular and molecular biology applications
Supplier: Thermo Scientific™ J66153MB
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
HPLC | |
Solid | |
beta-Bag Cell Peptide | |
RUO - research use only |
-30°C to -10°C | |
727.86 | |
25 mg | |
Arg-Leu-Arg-Phe-His |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
For Research Use Only
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'