Learn More
Alfa Aesar™ beta-Casomorphin (1-4), Amide, Bovine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66979MA
 View more versions of this product
Catalog No. AAJ66979MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
521.61
Bovine
Solid
RUO
beta-Casomorphin (1-4), amide, bovine
HPLC
10mg
Tyr-Pro-Phe-Pro-NH2
-30° to -10°C