Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ beta-Casomorphin, human, 95+%

Click to view available options
Quantity:
5mg
25mg
102029-74-3
C44H61N7O11
864.01
MFCD00076324
ADBHAJDGVKLXHK-LMXUZNBISA-N
h-tyr-pro-phe-val-glu-pro-ile-oh,beta casomorphin,b-casomorphin human,beta-casomorphin, human,beta-casomorphin-7 human,h-tyr-pro-phe-val-glu-proile-oh,l-isoleucine,l-tyrosyl-l-prolyl-l-phenylalanyl-l-valyl-l-a-glutamyl-l-prolyl,4s-4-2s-2-2s-2-2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl pyrrolidin-2-yl formamido-3-phenylpropanamido-3-methylbutanamido-5-2s-2-1s,2s-1-carboxy-2-methylbutyl carbamoyl pyrrolidin-1-yl-5-oxopentanoic acid,s-4-s-2-s-2-s-1-s-2-amino-3-4-hydroxyphenyl propanoyl pyrrolidine-2-carboxamido-3-phenylpropanamido-3-methylbutanamido-5-s-2-1s,2s-1-carboxy-2-methylbutyl carbamoyl pyrrolidin-1-yl-5-oxopentanoic acid
9832595
(2S,3S)-2-[[(2S)-1-[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidine-2-carbonyl]amino]-3-phenylpropanoyl]amino]-3-methylbutanoyl]amino]-4-carboxybutanoyl]pyrrolidine-2-carbonyl]amino]-3-methylpentanoic acid
CCC(C)C(C(=O)O)NC(=O)C1CCCN1C(=O)C(CCC(=O)O)NC(=O)C(C(C)C)NC(=O)C(CC2=CC=CC=C2)NC(=O)C3CCCN3C(=O)C(CC4=CC=C(C=C4)O)N
102029-74-3
C44H61N7O11
864.01
MFCD00076324
ADBHAJDGVKLXHK-LMXUZNBISA-N
h-tyr-pro-phe-val-glu-pro-ile-oh,beta casomorphin,b-casomorphin human,beta-casomorphin, human,beta-casomorphin-7 human,h-tyr-pro-phe-val-glu-proile-oh,l-isoleucine,l-tyrosyl-l-prolyl-l-phenylalanyl-l-valyl-l-a-glutamyl-l-prolyl,4s-4-2s-2-2s-2-2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl pyrrolidin-2-yl formamido-3-phenylpropanamido-3-methylbutanamido-5-2s-2-1s,2s-1-carboxy-2-methylbutyl carbamoyl pyrrolidin-1-yl-5-oxopentanoic acid,s-4-s-2-s-2-s-1-s-2-amino-3-4-hydroxyphenyl propanoyl pyrrolidine-2-carboxamido-3-phenylpropanamido-3-methylbutanamido-5-s-2-1s,2s-1-carboxy-2-methylbutyl carbamoyl pyrrolidin-1-yl-5-oxopentanoic acid
9832595
(2S,3S)-2-[[(2S)-1-[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidine-2-carbonyl]amino]-3-phenylpropanoyl]amino]-3-methylbutanoyl]amino]-4-carboxybutanoyl]pyrrolidine-2-carbonyl]amino]-3-methylpentanoic acid
CCC(C)C(C(=O)O)NC(=O)C1CCCN1C(=O)C(CCC(=O)O)NC(=O)C(C(C)C)NC(=O)C(CC2=CC=CC=C2)NC(=O)C3CCCN3C(=O)C(CC4=CC=C(C=C4)O)N

An opioid peptide

Quantity 5mg
Color White
Formula Weight 864.01
Physical Form Powder
Percent Purity ≥95%
Chemical Name or Material beta-Casomorphin, human

Recommended Storage : −30° to −10°C