Learn More
beta-Catenin Antibody (OTI9C1), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270508V
Catalog No. NBP270508V
Add to Cart
Description
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
beta-Catenin
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CTNNB1
Full length recombinant protein of human beta-Catenin (NP_001895) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Monkey
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI9C1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Mouse
Affinity Purified
Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
1499.0
Store at 4°C in the dark.
Recommendations
Recommendations