Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

beta-Catenin Mouse anti-Human, Mouse, Rat, Chicken, Primate, PE-Cyanine7, Clone: 12F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP154467PECY7

Catalog No. N154467PEC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat, Chicken, Primate samples. It is validated for Flow Cytometry.
Specifications

Specifications

beta-Catenin
12F7
Flow Cytometry
Conjugated, Purified
CTNNB1
Recombinant chicken beta Catenin fused to maltose binding protein. [UniProt No. O42486]
Protein G purified
Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
1499.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Chicken, Human, Mouse, Primate, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit