Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Catenin Mouse anti-Human, Rat, Monkey, Biotin, Clone: OTI12H7, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270509B
Description
beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
beta-Catenin | |
OTI12H7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
CTNNB1 | |
Full length recombinant protein of human beta-Catenin (NP_001895) produced in HEK293T cells. | |
Immunogen affinity purified | |
Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
Primary | |
1499.0 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Monkey, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title