Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin Mouse anti-Human, Rat, Monkey, PE/Atto594, Clone: OTI12H7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270509PEATT594

Catalog No. N270509P594


Add to cart

Description

Description

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

beta-Catenin
OTI12H7
Flow Cytometry
Affinity Purified, Conjugated
CTNNB1
Full length recombinant protein of human beta-Catenin (NP_001895) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
1499.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit