Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Catenin Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 3J7, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen beta Catenin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB056652MI
View Documents
MilliporeSigma
ZRB0566525UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB056654MI
View Documents
MilliporeSigma
ZRB056654X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

beta Catenin Monoclonal antibody specifically detects beta Catenin in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

beta Catenin
Unconjugated
Rabbit
Catenin beta-1
GST/His-tagged recombinant fragment corresponding to the first 100 amino acids from the N-terminal region of human beta-catenin.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
CTNNB1, CTNNB, OK/SW-cl.35, PRO2286
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit