Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin, Rabbit anti-Human, PE, Clone: CTNNB1/2030R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279888PE

Catalog No. NB011889


Add to cart

Description

Description

beta-Catenin Monoclonal specifically detects beta-Catenin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

beta-Catenin
Monoclonal
PE
PBS with 0.05% Sodium Azide
CTNNB1
Recombinant full-length human beta-Catenin protein
0.1 mL
Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Human
IgG
Flow Cytometry
CTNNB1/2030R
Flow Cytometry
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Rabbit
Protein A purified
RUO
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit