Learn More
Invitrogen™ beta Catenin (Total) Multispecies InstantOne™ ELISA Kit Non-distribution product as customer accommodation.
InstantOne ELISA
Manufacturer:  Invitrogen™ 858614111
Catalog No. 50-246-269
Add to cart
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
beta-Catenin (Total) Capture Antibody Reagent
beta-Catenin (Total) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate Seal
Description
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of human, mouse, and rat total beta-Catenin in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Beta-catenin, an adherens junction (AJ) protein, was originally identified as a component of cell-cell adhesion structures. AJs are necessary for the creation and maintenance of epithelial cell layers by regulating cell growth and adhesion between cells. Beta-catenin interacts with the cytoplasmic domain of E-cadherin and links E-cadherin to alpha-catenin, which in turn mediates anchorage of the E-cadherin complex to the cortical actin cytoskeleton. Studies show that Beta-catenin also binds to another cytoskeletal complex containing the adenomatous polyposis coli protein and microtubules, and interacts with several signaling pathways that include tyrosine kinases, phosphatases and Wnt/Wingless. The interplay between beta-catenin, cytoskeletal complexes and signaling pathways may regulate morphogenesis. Beta-catenin is expressed in several hair follicle cell types, basal and peripheral matrix cells, and cells of the outer and inner root sheats. A pathological role of beta-catenin has been identified in pilomatrixoma (PTR), medulloblastoma (MDB), colorectal cancer (CRC), ovarian cancer, and tumor development. In the nucleus, beta-catenin serves to co activate a family of Lef/Tcf transcription factors that stimulate transcription of target genes including those encoding cyclin D and c-myc that promote cell proliferation. The influence on cell proliferation is the molecular basis for the role of beta-catenin in tumorgenesis, specifically, solid tumors of the breast, colon, liver, lung, gastric, prostate, and skin.
Specifications
Specifications
P35222,Q02248,Q9WU82
ELISA
Capture Antibody
RUO
2°C to 8°C
1 hr. 10 min.
CTNNB,MRD19,armadillo
96 Tests
Cell Lysates
Colorimetric Microplate Reader
Biotin
12387, 1499, 84353
ELISA Kit
Cell Lysate,20 μL
1 hr. 20 min.
Human, Mouse, Rat
CTNNB1
Human, Mouse, Rat
HRP