Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ beta-D-Galactose pentaacetate, 98%

Manufacturer:  Alfa Aesar™ A1216322

Quantity 100g

 View more versions of this product

Catalog No. AAA1216322


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as a pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.

Notes
Store at room temperature. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

4163-60-4
390.34
98853
beta-D-Galactose pentaacetate
C16H22O11
MFCD00063259
1,2,3,4,6-Pentaacetyl-beta-D-galactose

Specifications

4163-60-4
390.34
MFCD00063259
100g
+24° (c=4 in chloroform)
98%
C16H22O11
142° to 146°C
98853
1,2,3,4,6-Pentaacetyl-beta-D-galactose
beta-D-Galactose pentaacetate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit