Alfa Aesar™ beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

Manufacturer: Alfa Aesar™ B2442606

Quantity 5g

 View more versions of this product

Catalog No. AAB2442606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13035-61-5
    Molecular Formula C13H18O9
    Formula Weight 318.28
    MDL Number MFCD00005358
    Beilstein 94078
    Synonym 1,2,3,5-Tetra-O-acetyl-beta-D-ribofuranose
    Chemical Name or Material beta-D-Ribofuranose 1,2,3,5-tetraacetate

    Specifications

    Formula Weight 318.28
    Molecular Formula C13H18O9
    Melting Point 80° to 83°C
    MDL Number MFCD00005358
    Quantity 5g
    Optical Rotation −13±0.5° (c=2.4 in Chloroform)
    CAS 13035-61-5
    Beilstein 94078
    Synonym 1,2,3,5-Tetra-O-acetyl-beta-D-ribofuranose
    Chemical Name or Material beta-D-Ribofuranose 1,2,3,5-tetraacetate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit