Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-endorphin Mouse anti-Human, Alexa Fluor™ 594, Clone: CLIP/1418, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP254334AF594

Catalog No. NP24334A94


Add to cart

Description

Description

Beta-endorphin Monoclonal antibody specifically detects Beta-endorphin in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Beta-endorphin
CLIP/1418
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
POMC
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF)
Protein A or G purified
Neuroscience, Nutrient Sensing in the Brain
Primary
5443.0
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit