Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-endorphin Mouse anti-Human, Clone: CLIP/1418, Novus Biologicals™

Mouse Monoclonal Antibody

$129.70 - $331.00

Specifications

Antigen Beta-endorphin
Clone CLIP/1418
Host Species Mouse
Isotype IgG1
Gene Alias ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25319220 View Documents Novus Biologicals
NBP25319220UG
20 ug Each for $129.70
Add to cart
 
NB253192100 View Documents Novus Biologicals
NBP253192100UG
100 ug Each for $331.00
Add to cart
 
NB253192200 View Documents Novus Biologicals
NBP253192200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Beta-endorphin Monoclonal antibody specifically detects Beta-endorphin in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page.
Specifications

Specifications

Beta-endorphin
Mouse
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
CLIP/1418
IgG1
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit