Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Galactosidase, anti-Escherichia coli, Clone: 13R4, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00146

Catalog No. 89-032-489


Add to cart

Specifications

Specifications

Beta-Galactosidase
13R4
Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG2c, lambda
RUO
Antibody
Monoclonal
E. Coli
Flow Cytometry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against beta-galactosidase.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
This antibody recognizes wild-type E. coli beta-galactosidase.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit