Learn More
Beta-galactosidase, Clone: 13R4, Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00045
Catalog No. 89-032-388
Add to cart
Specifications
Specifications
Beta-galactosidase
13R4
Flow Cytometry (1 ug/mL) Negative control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG1, lambda
RUO
Antibody
Monoclonal
E. Coli
Flow Cytometry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against beta-galactosidase.
200 μg
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
This antibody recognizes wild-type E. coli beta-galactosidase.