Learn More
beta Galactosidase Mouse anti-E. coli, Alexa Fluor™ 532, Clone: 13R4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252702AF532
Catalog No. NP22702A32
Add to cart
Description
Description
beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
beta Galactosidase
13R4
Western Blot, Flow Cytometry
EC 3.2.1.23; b-gal
Beta-galactosidase.
0.1 mL
Primary
13R4 recognizes wild-type E. coli beta-galactosidase.
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Lambda
Store at 4°C in the dark.
Monoclonal
E. coli